Assay Details
Gene Symbol: PRSS55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607709.3 | XP_005607766.1 | 340 | 106 | ||
XM_023636249.1 | XP_023492017.1 | 340 | 106 |

More Information
Set Membership: |
![]() ![]() |
