Assay Details
Gene Symbol: SAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636280.1 | XP_023492048.1 | 2020 | 72 | ||
XM_023636281.1 | XP_023492049.1 | 2020 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
