Assay Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607723.3 | XP_005607780.1 | 614 | 107 | ||
XM_005607724.3 | XP_005607781.1 | 789 | 107 | |||
XM_005607725.3 | XP_005607782.1 | 623 | 107 | |||
XM_023636300.1 | XP_023492068.1 | 3440 | 107 | |||
XM_023636301.1 | XP_023492069.1 | 805 | 107 | |||
XM_023636302.1 | XP_023492070.1 | 3339 | 107 | |||
XM_023636303.1 | XP_023492071.1 | 695 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
