Assay Details
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499669.6 | XP_001499719.1 | 745 | 88 | ||
XM_005607741.3 | XP_005607798.1 | 842 | 88 | |||
XM_005607742.3 | XP_005607799.1 | 745 | 88 | |||
GenBank mRNA | HQ890057.1 | - | 1 - 2 | 103 | 88 | |
JL641412.1 | - | 1841 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
