Assay Details
Gene Symbol: NPY1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499988.5 | XP_001500038.1 | 1038 | 98 | ||
XM_005607751.3 | XP_005607808.1 | 965 | 98 | |||
XM_005607752.3 | XP_005607809.1 | 1147 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
