Assay Details
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500334.5 | XP_001500384.1 | 994 | 76 | ||
GenBank mRNA | JL616521.1 | - | 7 - 8 | 950 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
