Assay Details
Gene Symbol: GUCY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636397.1 | XP_023492165.1 | 1899 | 80 | ||
XM_023636398.1 | XP_023492166.1 | 2067 | 80 | |||
XM_023636399.1 | XP_023492167.1 | 1844 | 80 | |||
XM_023636401.1 | XP_023492169.1 | 1919 | 80 | |||
XM_023636402.1 | XP_023492170.1 | 1907 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
