Assay Details
Gene Symbol: RNF175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737991.2 | XP_014593477.2 | 865 | 91 | ||
XM_023636415.1 | XP_023492183.1 | 868 | 91 | |||
XM_023636416.1 | XP_023492184.1 | 878 | 91 | |||
XM_023636417.1 | XP_023492185.1 | 878 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
