Assay Details
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636454.1 | XP_023492222.1 | 3312 | 83 | ||
XM_023636455.1 | XP_023492223.1 | 3297 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
