Assay Details
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607826.3 | XP_005607883.1 | 477 | 91 | ||
XM_005607827.3 | XP_005607884.1 | 484 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
