Assay Details
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636548.1 | XP_023492316.1 | 4626 | 94 | ||
XM_023636549.1 | XP_023492317.1 | 4626 | 94 | |||
XM_023636550.1 | XP_023492318.1 | 4626 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
