Assay Details
Gene Symbol: TNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737997.2 | XP_014593483.2 | 419 | 93 | ||
XM_023636623.1 | XP_023492391.1 | 376 | 93 | |||
XM_023636624.1 | XP_023492392.1 | 290 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
