Assay Details
Gene Symbol: UGT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636653.1 | XP_023492421.1 | 1368 | 81 | ||
XM_023636654.1 | XP_023492422.1 | 1330 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
