Assay Details
Gene Symbol: ELOVL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005607891.3 | XP_005607948.1 | 357 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
