Assay Details
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636752.1 | XP_023492520.1 | 676 | 92 | ||
XM_023636753.1 | XP_023492521.1 | 686 | 92 | |||
GenBank mRNA | JL640794.1 | - | 5 - 6 | 613 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
