Assay Details
Gene Symbol: ARHGEF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503179.6 | XP_001503229.4 | 2317 | 81 | ||
XM_023636784.1 | XP_023492552.1 | 2299 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
