Assay Details
Gene Symbol: SERPINB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734698.2 | XP_014590184.1 | 954 | 73 | ||
XM_014734699.2 | XP_014590185.1 | 933 | 73 | |||
XM_023624361.1 | XP_023480129.1 | 954 | 73 | |||
XM_023624362.1 | XP_023480130.1 | 954 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
