Assay Details
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624393.1 | XP_023480161.1 | 1835 | 62 | ||
XM_023624395.1 | XP_023480163.1 | 1866 | 62 | |||
XM_023624396.1 | XP_023480164.1 | 1559 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
