Assay Details
Gene Symbol: ELOVL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624455.1 | XP_023480223.1 | 868 | 104 | ||
XM_023624456.1 | XP_023480224.1 | 778 | 104 | |||
XM_023624457.1 | XP_023480225.1 | 727 | 104 | |||
XM_023624458.1 | XP_023480226.1 | 2175 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
