Assay Details
Gene Symbol: NOL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915240.3 | XP_001915275.1 | 468 | 101 | ||
GenBank mRNA | JL622039.1 | - | 1 - 2 | 87 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
