Assay Details
Gene Symbol: ACOT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734221.1 | XP_014589707.1 | 228 | 88 | ||
XM_023624530.1 | XP_023480298.1 | 402 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
