Assay Details
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603640.3 | XP_005603697.1 | 934 | 90 | ||
XM_023624111.1 | XP_023479879.1 | 910 | 90 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603640.3 | XP_005603697.1 | 934 | 90 | ||
XM_023624111.1 | XP_023479879.1 | 910 | 90 |