Assay Details
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490108.6 | XP_001490158.2 | 1010 | 88 | ||
XM_023624232.1 | XP_023480000.1 | 276 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
