Assay Details
Gene Symbol: EGFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493316.6 | XP_001493366.3 | 804 | 140 | ||
XM_005603766.2 | XP_005603823.1 | 738 | 140 | |||
XM_023624652.1 | XP_023480420.1 | 747 | 140 | |||
XM_023624653.1 | XP_023480421.1 | 747 | 140 | |||
XM_023624654.1 | XP_023480422.1 | 804 | 140 | |||
XM_023624655.1 | XP_023480423.1 | 1266 | 140 | |||
XM_023624656.1 | XP_023480424.1 | 1172 | 140 | |||
XM_023624657.1 | XP_023480425.1 | 888 | 140 | |||
XM_023624658.1 | XP_023480426.1 | 855 | 140 | |||
XM_023624659.1 | XP_023480427.1 | 1244 | 140 | |||
XM_023624660.1 | XP_023480428.1 | 842 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
