Assay Details
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603819.3 | XP_005603876.1 | 633 | 84 | ||
XM_023624729.1 | XP_023480497.1 | 757 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
