Assay Details
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624748.1 | XP_023480516.1 | 532 | 102 | ||
XM_023624749.1 | XP_023480517.1 | 851 | 102 | |||
XM_023624750.1 | XP_023480518.1 | 498 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
