Assay Details
Gene Symbol: TEAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624804.1 | XP_023480572.1 | 1488 | 85 | ||
XM_023624805.1 | XP_023480573.1 | 1322 | 85 | |||
XM_023624806.1 | XP_023480574.1 | 1456 | 85 | |||
XM_023624807.1 | XP_023480575.1 | 1423 | 85 | |||
XM_023624808.1 | XP_023480576.1 | 1424 | 85 | |||
XM_023624809.1 | XP_023480577.1 | 1492 | 85 | |||
XM_023624810.1 | XP_023480578.1 | 1427 | 85 | |||
XM_023624811.1 | XP_023480579.1 | 1429 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
