Assay Details
Gene Symbol: TREML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496531.4 | XP_001496581.1 | 74 | 64 | ||
XM_023624903.1 | XP_023480671.1 | 74 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
