Assay Details
Gene Symbol: TREML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496531.4 | XP_001496581.1 | 608 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
