Assay Details
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603890.3 | XP_005603947.1 | 158 | 75 | ||
XM_005603891.3 | XP_005603948.1 | 159 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
