Assay Details
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501311.5 | XP_001501361.1 | 265 | 80 | ||
XM_005603912.3 | XP_005603969.1 | 263 | 80 | |||
XM_014734413.2 | XP_014589899.1 | 782 | 80 | |||
XM_014734414.2 | XP_014589900.1 | 771 | 80 | |||
XM_014734415.2 | XP_014589901.1 | 308 | 80 | |||
GenBank mRNA | JL616010.2 | - | 2 - 3 | 134 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
