Assay Details
Gene Symbol: BICRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734429.2 | XP_014589915.1 | 2241 | 62 | ||
XM_023624943.1 | XP_023480711.1 | 2826 | 62 | |||
XM_023624944.1 | XP_023480712.1 | 2593 | 62 |

More Information
Set Membership: |
![]() ![]() |
