Assay Details
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502138.5 | XP_001502188.1 | 408 | 81 | ||
XM_023625008.1 | XP_023480776.1 | 718 | 81 | |||
XM_023625009.1 | XP_023480777.1 | 610 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
