Assay Details
Gene Symbol: MRPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502257.5 | XP_001502307.1 | 205 | 79 | ||
XM_005603958.3 | XP_005604015.1 | 218 | 79 | |||
XM_014734453.2 | XP_014589939.1 | 337 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
