Assay Details
Gene Symbol: LOC100068322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498174.4 | XP_001498224.1 | 1017 | 144 | ||
XM_014734472.2 | XP_014589958.1 | 1019 | 144 |

More Information
Set Membership: |
![]() ![]() |
