Assay Details
Gene Symbol: ADGRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502766.3 | XP_001502816.2 | 424 | 66 | ||
XM_014734483.2 | XP_014589969.2 | 425 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
