Assay Details
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003363723.4 | XP_003363771.1 | 443 | 61 | ||
XM_014734489.2 | XP_014589975.2 | 299 | 61 | |||
XM_023625080.1 | XP_023480848.1 | 385 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
