Assay Details
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001918274.4 | XP_001918309.1 | 1938 | 60 | ||
XM_023625092.1 | XP_023480860.1 | 2033 | 60 | |||
XM_023625093.1 | XP_023480861.1 | 1939 | 60 | |||
XM_023625094.1 | XP_023480862.1 | 1439 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
