Assay Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005603993.3 | XP_005604050.1 | 2637 | 79 | ||
XM_005603994.3 | XP_005604051.1 | 2565 | 79 | |||
XM_023625105.1 | XP_023480873.1 | 2595 | 79 | |||
XM_023625106.1 | XP_023480874.1 | 2571 | 79 | |||
XM_023625107.1 | XP_023480875.1 | 2595 | 79 | |||
XM_023625108.1 | XP_023480876.1 | 2511 | 79 | |||
XM_023625109.1 | XP_023480877.1 | 2445 | 79 | |||
XM_023625110.1 | XP_023480878.1 | 2439 | 79 | |||
XM_023625112.1 | XP_023480880.1 | 2595 | 79 | |||
XM_023625113.1 | XP_023480881.1 | 2385 | 79 | |||
XM_023625114.1 | XP_023480882.1 | 2595 | 79 | |||
XM_023625115.1 | XP_023480883.1 | 2366 | 79 | |||
XM_023625116.1 | XP_023480884.1 | 1020 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
