Assay Details
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625125.1 | XP_023480893.1 | 843 | 58 | ||
XM_023625126.1 | XP_023480894.1 | 837 | 58 | |||
XM_023625127.1 | XP_023480895.1 | 866 | 58 | |||
GenBank mRNA | JL616014.2 | - | 2 - 3 | 441 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
