Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001914799.5 | XP_001914834.1 | 450 | 81 | ||
XM_023625365.1 | XP_023481133.1 | 454 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
