Assay Details
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625389.1 | XP_023481157.1 | 456 | 55 | ||
XM_023625390.1 | XP_023481158.1 | 291 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
