Assay Details
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495070.4 | XP_001495120.1 | 513 | 105 | ||
XM_005604243.3 | XP_005604300.1 | 732 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
