Assay Details
Gene Symbol: PLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493894.6 | XP_001493944.5 | 929 | 76 | ||
XM_023625668.1 | XP_023481436.1 | 939 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
