Assay Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496899.6 | XP_001496949.2 | 448 | 83 | ||
XM_005604269.3 | XP_005604326.1 | 209 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604269.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
