Assay Details
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734882.2 | XP_014590368.2 | 139 | 64 | ||
XM_023625672.1 | XP_023481440.1 | 139 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
