Assay Details
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604274.3 | XP_005604331.1 | 540 | 68 | ||
XM_005604275.3 | XP_005604332.1 | 585 | 68 | |||
XM_005604276.3 | XP_005604333.1 | 510 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
