Assay Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500213.6 | XP_001500263.1 | 572 | 76 | ||
XM_003363859.4 | XP_003363907.1 | 831 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
