Assay Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625791.1 | XP_023481559.1 | 322 | 66 | ||
XM_023625792.1 | XP_023481560.1 | 450 | 66 | |||
GenBank mRNA | HQ890101.1 | - | 1 - 2 | 95 | 66 | |
JL641531.1 | - | 1 - 2 | 110 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
