Assay Details
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625847.1 | XP_023481615.1 | 592 | 85 | ||
XM_023625848.1 | XP_023481616.1 | 591 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
